Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-chloro-4-methyl-7-hydroxycoumarin, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass Bottle
6174-86-3
C10H7ClO3
210.62
MFCD00006851
ODZHLDRQCZXQFQ-UHFFFAOYSA-N
chlorferone,3-chloro-7-hydroxy-4-methyl-2h-chromen-2-one,3-chloro-4-methyl-7-hydroxycoumarin,chlorferron,3-chloro-7-hydroxy-4-methylcoumarin,7-hydroxy-4-methyl-3-chlorocoumarin,chlorferon,3-chloro-4-methylumbelliferone,3-chloro-7-hydroxy-4-methyl-2-benzopyrone,2h-1-benzopyran-2-one, 3-chloro-7-hydroxy-4-methyl
5355079
CHEBI:38620
3-chloro-7-hydroxy-4-methylchromen-2-one
CC1=C(C(=O)OC2=C1C=CC(=C2)O)Cl
6174-86-3
C10H7ClO3
210.62
MFCD00006851
ODZHLDRQCZXQFQ-UHFFFAOYSA-N
chlorferone,3-chloro-7-hydroxy-4-methyl-2h-chromen-2-one,3-chloro-4-methyl-7-hydroxycoumarin,chlorferron,3-chloro-7-hydroxy-4-methylcoumarin,7-hydroxy-4-methyl-3-chlorocoumarin,chlorferon,3-chloro-4-methylumbelliferone,3-chloro-7-hydroxy-4-methyl-2-benzopyrone,2h-1-benzopyran-2-one, 3-chloro-7-hydroxy-4-methyl
5355079
CHEBI:38620
3-chloro-7-hydroxy-4-methylchromen-2-one
CC1=C(C(=O)OC2=C1C=CC(=C2)O)Cl
Packaging Glass Bottle
Color White
Quantity 5g
Melting Point 246.0°C to 247.0°C
Infrared Spectrum Authentic
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (HPLC)
Formula Weight 210.62
Physical Form Powder
Percent Purity 97%
Chemical Name or Material 3-chloro-4-methyl-7-hydroxycoumarin
Show More Show Less

GHS Signal Word: Warning

EINECSNumber : 228-217-8