Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Chloro-4-methylaniline, 98%

Click to view available options
Quantity:
250g
1000g
5000g
95-74-9
C7H8ClN
141.598
MFCD00007773
RQKFYFNZSHWXAW-UHFFFAOYSA-N
3-chloro-p-toluidine,2-chloro-4-aminotoluene,4-amino-2-chlorotoluene,benzenamine, 3-chloro-4-methyl,3-chloro-4-methylbenzenamine,3-chloro-4-methylphenylamine,1-amino-3-chloro-4-methylbenzene,4-methyl-3-chloroaniline,p-toluidine, 3-chloro,gull toxicant
7255
CHEBI:37824
3-chloro-4-methylaniline
CC1=C(C=C(C=C1)N)Cl
95-74-9
C7H8ClN
141.598
MFCD00007773
RQKFYFNZSHWXAW-UHFFFAOYSA-N
3-chloro-p-toluidine,2-chloro-4-aminotoluene,4-amino-2-chlorotoluene,benzenamine, 3-chloro-4-methyl,3-chloro-4-methylbenzenamine,3-chloro-4-methylphenylamine,1-amino-3-chloro-4-methylbenzene,4-methyl-3-chloroaniline,p-toluidine, 3-chloro,gull toxicant
7255
CHEBI:37824
3-chloro-4-methylaniline
CC1=C(C=C(C=C1)N)Cl
Quantity 1000g
Density 1.18
Refractive Index 1.584
Melting Point 22°C to 24°C
Boiling Point 236°C to 238°C
Flash Point 130°C (266°F)
UN Number UN2239
Beilstein 636511
Formula Weight 141.6
Percent Purity 98%
Chemical Name or Material 3-Chloro-4-methylaniline
Show More Show Less

GHS H Statement
H331-H373-H302-H312-H315-H319
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.

P261-P264b-P270-P272-P280-P301+P312-P302+P352-P305+P351+P338-P312-P330-P333+P313-P361-P363-P501c

H302-H311-H315-H317-H319

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: CHLOROTOLUIDINES, SOLID

EINECSNumber : 202-446-3

RTECSNumber : XU5111000

TSCA : Yes

Recommended Storage : Ambient temperatures