Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Chloro-4-methylaniline 99%, ACROS Organics™

Click to view available options
Quantity:
250g
1kg
Packaging:
Glass bottle
95-74-9
C7H8ClN
141.598
RQKFYFNZSHWXAW-UHFFFAOYSA-N
3-chloro-p-toluidine,2-chloro-4-aminotoluene,4-amino-2-chlorotoluene,benzenamine, 3-chloro-4-methyl,3-chloro-4-methylbenzenamine,3-chloro-4-methylphenylamine,1-amino-3-chloro-4-methylbenzene,4-methyl-3-chloroaniline,p-toluidine, 3-chloro,gull toxicant
7255
CHEBI:37824
3-chloro-4-methylaniline
CC1=C(C=C(C=C1)N)Cl
95-74-9
C7H8ClN
141.598
RQKFYFNZSHWXAW-UHFFFAOYSA-N
3-chloro-p-toluidine,2-chloro-4-aminotoluene,4-amino-2-chlorotoluene,benzenamine, 3-chloro-4-methyl,3-chloro-4-methylbenzenamine,3-chloro-4-methylphenylamine,1-amino-3-chloro-4-methylbenzene,4-methyl-3-chloroaniline,p-toluidine, 3-chloro,gull toxicant
7255
CHEBI:37824
3-chloro-4-methylaniline
CC1=C(C=C(C=C1)N)Cl
Density 1.1700g/mL
Packaging Glass bottle
Quantity 1kg
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula ClC6H3(CH3)NH2
Beilstein 12, 988
Formula Weight 141.6
Chemical Name or Material 3-Chloro-4-methylaniline, 99%

GHS P Statement
Harmful if swallowed.
Toxic in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
Very toxic to aquatic life with long lasting effects.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Wash face, hands and any exposed skin thoroughly after handling.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 202-446-3

RTECSNumber : XU5111000

TSCA : TSCA