Alfa Aesar™ 3-Chloro-4-methylaniline hydrochloride, 97%

Manufacturer: Alfa Aesar™ H3134214

Quantity 25g

 View more versions of this product

Catalog No. AAH3134214

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7745-89-3
    Molecular Formula C7H8ClN·HCl
    Formula Weight 178.06
    MDL Number MFCD00242925
    Chemical Name or Material 3-Chloro-4-methylaniline hydrochloride

    Specifications

    Formula Weight 178.06
    Molecular Formula C7H8ClN·HCl
    MDL Number MFCD00242925
    CAS 7745-89-3
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H373-H302-H312-H332-H315-H319-H335
    May cause damage to organs through prolonged or repeated exposure.
    Harmful if swallowed.
    Harmful in contact with skin.
    Harmful if inhaled.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P280h-P305+P351+P338
    Wear protective gloves/clothing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    RTECS Number XU5425000
    Melting Point ≈260°C (subl.)
    Quantity 25g
    Chemical Name or Material 3-Chloro-4-methylaniline hydrochloride
    Assay 97%

    GHS H Statement
    H373-H302-H312-H332-H315-H319-H335
    May cause damage to organs through prolonged or repeated exposure.
    Harmful if swallowed.
    Harmful in contact with skin.
    Harmful if inhaled.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P280h-P305+P351+P338
    Wear protective gloves/clothing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit