Alfa Aesar™ 3-Chloro-4-methylphenol, 97%

Manufacturer: Alfa Aesar™ H2738514

Quantity 25g

 View more versions of this product

Catalog No. AAH2738514

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 615-62-3
    Molecular Formula C7H7ClO
    Formula Weight 142.59
    MDL Number MFCD00060319
    UN Number UN3437
    Synonym 3-Chloro-p-cresol
    Chemical Name or Material 3-Chloro-4-methylphenol

    Specifications

    Formula Weight 142.59
    Molecular Formula C7H7ClO
    MDL Number MFCD00060319
    Synonym 3-Chloro-p-cresol
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H301-H311-H331-H318-H315-H335
    Toxic if swallowed.
    Toxic in contact with skin.
    Toxic if inhaled.
    Causes serious eye damage.
    Causes skin irritation.
    May cause respiratory irritation.
    Quantity 25g
    TSCA TSCA
    CAS 615-62-3
    UN Number UN3437
    Health Hazard 3 GHS P Statement
    P261-P301+P310-P305+P351+P338-P361-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: CHLOROCRESOLS, SOLID
    Boiling Point 228°C
    Melting Point 51° to 53°C
    Chemical Name or Material 3-Chloro-4-methylphenol
    Assay 97%

    GHS H Statement
    H301-H311-H331-H318-H315-H335
    Toxic if swallowed.
    Toxic in contact with skin.
    Toxic if inhaled.
    Causes serious eye damage.
    Causes skin irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P301+P310-P305+P351+P338-P361-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit