Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-chloro-4-methylphenyl isocyanate, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
Packaging:
Glass bottle
28479-22-3
C8H6ClNO
167.59
UKTKKMZDESVUEE-UHFFFAOYSA-N
3-chloro-4-methylphenyl isocyanate,3-chloro-4-methylphenylisocyanate,benzene, 2-chloro-4-isocyanato-1-methyl,3-chloro-p-tolyl isocyanate,isocyanic acid 3-chloro-4-methylphenyl ester,3-chloro-4-methyl phenyl isocyanate,acmc-1ar7g,3-chlor-4-methylphenylisocyanat,2-chloro-4-isocyanato toluene,3-chloro-4-methylphenyl isocyanate un2236 poison
62832
2-chloro-4-isocyanato-1-methylbenzene
CC1=C(C=C(C=C1)N=C=O)Cl
28479-22-3
C8H6ClNO
167.59
UKTKKMZDESVUEE-UHFFFAOYSA-N
3-chloro-4-methylphenyl isocyanate,3-chloro-4-methylphenylisocyanate,benzene, 2-chloro-4-isocyanato-1-methyl,3-chloro-p-tolyl isocyanate,isocyanic acid 3-chloro-4-methylphenyl ester,3-chloro-4-methyl phenyl isocyanate,acmc-1ar7g,3-chlor-4-methylphenylisocyanat,2-chloro-4-isocyanato toluene,3-chloro-4-methylphenyl isocyanate un2236 poison
62832
2-chloro-4-isocyanato-1-methylbenzene
CC1=C(C=C(C=C1)N=C=O)Cl
Specific Gravity 1.23
Packaging Glass bottle
Refractive Index 1.5560 to 1.5580 (20°C, 589nm)
Density 1.2300g/mL
Color Colorless to Yellow
Quantity 5g
Flash Point 110°C
Infrared Spectrum Authentic
Boiling Point 225.0°C
Melting Point 23.0°C
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (GC)
Solubility Information Solubility in water: decomposes
Formula Weight 167.59
Physical Form Liquid or Low Melting Solid
Percent Purity 98%
Chemical Name or Material 3-Chloro-4-methylphenyl isocyanate
Show More Show Less

GHS H Statement
Fatal if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes severe skin burns and eye damage.
Harmful to aquatic life with long lasting effects.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Do not breat

GHS Signal Word: Danger

EINECSNumber : 249-050-7