Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Chloro-4-nitrotoluene, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
38939-88-7
C7H6ClNO2
171.58
MFCD02683043
KGSQRFPDZCBVBS-UHFFFAOYSA-N
3-chloro-4-nitrotoluene,2-chloro-4-methyl-1-nitro-benzene,benzene, 2-chloro-4-methyl-1-nitro,pubchem4534,acmc-1afh8,ksc493s6j
123478
2-chloro-4-methyl-1-nitrobenzene
CC1=CC(=C(C=C1)[N+](=O)[O-])Cl
38939-88-7
C7H6ClNO2
171.58
MFCD02683043
KGSQRFPDZCBVBS-UHFFFAOYSA-N
3-chloro-4-nitrotoluene,2-chloro-4-methyl-1-nitro-benzene,benzene, 2-chloro-4-methyl-1-nitro,pubchem4534,acmc-1afh8,ksc493s6j
123478
2-chloro-4-methyl-1-nitrobenzene
CC1=CC(=C(C=C1)[N+](=O)[O-])Cl
Refractive Index 1.5610 to 1.5640
Infrared Spectrum Authentic
Melting Point 22.0°C
Packaging Glass bottle
Quantity 5g
Boiling Point 108.0°C (3.0 mmHg)
Color Yellow
Flash Point >230°C
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (GC)
Linear Formula CH3C6H3(NO2)Cl
Beilstein 05, IV, 856
Solubility Information Solubility in water: insoluble.
Formula Weight 171.58
Physical Form Liquid After Melting
Percent Purity 98%
Chemical Name or Material 3-Chloro-4-nitrotoluene
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Harmful if inhaled.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 254-199-6

TSCA : TSCA