Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Chloro-4-(trifluoromethoxy)aniline, 97%

Click to view available options
Quantity:
2g
10g
64628-73-5
C7H5ClF3NO
211.568
MFCD00190126
ZPKUUNGPBSRPRM-UHFFFAOYSA-N
3-chloro-4-trifluoromethoxy aniline,benzenamine, 3-chloro-4-trifluoromethoxy,3-chloro-4-trifluoromethoxy phenylamine,pubchem2949,acmc-1b4hb,ksc496e4r,3-chloro4-trifluoromethoxyaniline,3-chloro-4-trifluoromethoxy-aniline,3-chloro-4-trifluoromethoxy-phenylamine
2773843
3-chloro-4-(trifluoromethoxy)aniline
C1=CC(=C(C=C1N)Cl)OC(F)(F)F
64628-73-5
C7H5ClF3NO
211.568
MFCD00190126
ZPKUUNGPBSRPRM-UHFFFAOYSA-N
3-chloro-4-trifluoromethoxy aniline,benzenamine, 3-chloro-4-trifluoromethoxy,3-chloro-4-trifluoromethoxy phenylamine,pubchem2949,acmc-1b4hb,ksc496e4r,3-chloro4-trifluoromethoxyaniline,3-chloro-4-trifluoromethoxy-aniline,3-chloro-4-trifluoromethoxy-phenylamine
2773843
3-chloro-4-(trifluoromethoxy)aniline
C1=CC(=C(C=C1N)Cl)OC(F)(F)F
Applications
3-Chloro-4-(trifluoromethoxy)aniline is used as a primary and secondary intermediate in chemical synthesis.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible material is strong oxidizing agents.
Quantity 2g
Melting Point 29°C to 30°C
Beilstein 6314199
Formula Weight 211.57
Percent Purity 97%
Chemical Name or Material 3-Chloro-4-(trifluoromethoxy)aniline

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger