Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Chloro-4-(trifluoromethoxy)benzonitrile, 97+%

Click to view available options
Quantity:
1g
5g
129604-26-8
C8H3ClF3NO
221.563
MFCD01631557
ITKCOGUNHYXLND-UHFFFAOYSA-N
3-chloro-4-trifluoromethoxy benzonitrile,3-chloro-4-trifluoromethoxy-benzonitrile,benzonitrile, 3-chloro-4-trifluoromethoxy,3-chloro-4-trifluoromethoxy benzenecarbonitrile,pubchem10175,acmc-20a6wk,ksc496g4n
2773846
3-chloro-4-(trifluoromethoxy)benzonitrile
C1=CC(=C(C=C1C#N)Cl)OC(F)(F)F
129604-26-8
C8H3ClF3NO
221.563
MFCD01631557
ITKCOGUNHYXLND-UHFFFAOYSA-N
3-chloro-4-trifluoromethoxy benzonitrile,3-chloro-4-trifluoromethoxy-benzonitrile,benzonitrile, 3-chloro-4-trifluoromethoxy,3-chloro-4-trifluoromethoxy benzenecarbonitrile,pubchem10175,acmc-20a6wk,ksc496g4n
2773846
3-chloro-4-(trifluoromethoxy)benzonitrile
C1=CC(=C(C=C1C#N)Cl)OC(F)(F)F
Quantity 5g
Melting Point 39°C to 41°C
UN Number UN3439
Formula Weight 221.57
Percent Purity ≥97%
Chemical Name or Material 3-Chloro-4-(trifluoromethoxy)benzonitrile

GHS H Statement
H311-H302-H332-H315-H319
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P280h-P305+P351+P338-P309-P310
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.