Alfa Aesar™ 3-Chloro-4-(trifluoromethyl)benzaldehyde, 97%

Manufacturer: Alfa Aesar™ H3163503

Quantity 1g

 View more versions of this product

Catalog No. AAH3163503

  Check Availability
Add to Cart

    Chemical Identifiers

    Molecular Formula C8H4ClF3O
    Formula Weight 208.57
    MDL Number MFCD03095197
    Chemical Name or Material 3-Chloro-4-(trifluoromethyl)benzaldehyde

    Specifications

    Formula Weight 208.57
    MDL Number MFCD03095197
    Quantity 1g
    Molecular Formula C8H4ClF3O
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H315-H319
    Causes skin irritation.
    Causes serious eye irritation.
    Health Hazard 3 GHS P Statement
    P280g-P305+P351+P338
    Wear protective gloves.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Chemical Name or Material 3-Chloro-4-(trifluoromethyl)benzaldehyde
    Assay 97%

    Description

    Applications
    3-Chloro-4-(trifluoromethyl)benzaldehyde is used in synthesis of organic compunds in γ-Aminobutyrate-A Receptor Modulation by 3-Aryl-1-(arylsulfonyl)- 1,4,5,6-tetrahydropyridazines.

    Solubility
    Insoluble in water.

    Notes
    Air Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air.

    GHS H Statement
    H315-H319
    Causes skin irritation.
    Causes serious eye irritation.

    GHS P Statement
    P280g-P305+P351+P338
    Wear protective gloves.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit