Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Chloro-4-(trifluoromethylthio)aniline, 97%

Click to view available options
Quantity:
1g
5g
64628-74-6
C7H5ClF3NS
227.629
MFCD00190131
BLOPJEJSRKMVSU-UHFFFAOYSA-N
3-chloro-4-trifluoromethylthio aniline,3-chloro-4-trifluoromethyl thio aniline,3-chloro-4-trifluoromethyl sulfanyl aniline,benzenamine,3-chloro-4-trifluoromethyl thio,3-chloro-4-trifluoromethylthio phenylamine,pubchem2950,3-chloro-4-trifluoromethylthioaniline,3-chloro-4-trifluoromethylsulfanyl aniline,3-chloranyl-4-trifluoromethylsulfanyl aniline,3-chloro-4-trifluoromethylsulfanyl-phenylamine
2736713
3-chloro-4-(trifluoromethylsulfanyl)aniline
C1=CC(=C(C=C1N)Cl)SC(F)(F)F
64628-74-6
C7H5ClF3NS
227.629
MFCD00190131
BLOPJEJSRKMVSU-UHFFFAOYSA-N
3-chloro-4-trifluoromethylthio aniline,3-chloro-4-trifluoromethyl thio aniline,3-chloro-4-trifluoromethyl sulfanyl aniline,benzenamine,3-chloro-4-trifluoromethyl thio,3-chloro-4-trifluoromethylthio phenylamine,pubchem2950,3-chloro-4-trifluoromethylthioaniline,3-chloro-4-trifluoromethylsulfanyl aniline,3-chloranyl-4-trifluoromethylsulfanyl aniline,3-chloro-4-trifluoromethylsulfanyl-phenylamine
2736713
3-chloro-4-(trifluoromethylsulfanyl)aniline
C1=CC(=C(C=C1N)Cl)SC(F)(F)F
Melting Point 41°C to 44°C
Quantity 1g
UN Number UN2811
Beilstein 6314200
Formula Weight 227.64
Percent Purity 97%
Chemical Name or Material 3-Chloro-4-(trifluoromethylthio)aniline

GHS H Statement
H311-H302-H332-H315-H319
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P280h-P305+P351+P338-P309-P310
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.