Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-chloro-5-fluoroaniline, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
4863-91-6
C6H5ClFN
145.56
MFCD03407962
LPIFAHAICWJMRR-UHFFFAOYSA-N
pubchem2953,acmc-209kdj,3-chloro-5-fluorobenzenamine,5-chloro-3-fluorophenylamine,ksc493m9f,3-chloro-5-fluoro-phenylamine,buttpark 44\01-96
2734838
3-chloro-5-fluoroaniline
C1=C(C=C(C=C1F)Cl)N
4863-91-6
C6H5ClFN
145.56
MFCD03407962
LPIFAHAICWJMRR-UHFFFAOYSA-N
pubchem2953,acmc-209kdj,3-chloro-5-fluorobenzenamine,5-chloro-3-fluorophenylamine,ksc493m9f,3-chloro-5-fluoro-phenylamine,buttpark 44\01-96
2734838
3-chloro-5-fluoroaniline
C1=C(C=C(C=C1F)Cl)N
Specific Gravity 1.45
Density 1.4500g/mL
Refractive Index 1.5610 to 1.563
Infrared Spectrum Authentic
Quantity 25g
Packaging Glass bottle
Color Orange to Yellow
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (GC)
Linear Formula ClC6H3(F)NH2
Solubility Information Solubility in water: soluble
Formula Weight 145.56
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material 3-chloro-5-fluoroaniline
Show More Show Less

GHS H Statement
Harmful in contact with skin.
Causes serious eye irritation.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
Harmful if inhaled.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor

GHS Signal Word: Warning

Recommended Storage : Darkens on exposure to light and air