Alfa Aesar™ 3-Chloro-5-fluoroanisole, 97%

Manufacturer: Alfa Aesar™ H5526614

Quantity 25g

 View more versions of this product

Catalog No. AAH5526614

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 202925-08-4
    Formula Weight 160.58
    MDL Number MFCD00070770
    Chemical Name or Material 3-Chloro-5-fluoroanisole

    Specifications

    Formula Weight 160.58
    MDL Number MFCD00070770
    Refractive Index 1.5110
    Physical Form Liquid
    CAS 202925-08-4
    Density 1.265g/mL
    Boiling Point 189°C
    Color Colorless to yellow
    Quantity 25g
    Chemical Name or Material 3-Chloro-5-fluoroanisole
    Assay 97%

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit