Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Chloro-5-fluorobenzoic acid, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
25026-64-6
C7H4ClFO2
174.56
MFCD00973904
QFMRANWPGGSNHS-UHFFFAOYSA-N
3-chloro-5-fluoro-benzoic acid,3-fluoro-5-chlorobenzoic acid,5-chloro-3-fluorobenzoic acid,benzoic acid, 3-chloro-5-fluoro,3-chloro-5-fluorobenzoicacid,pubchem1373,acmc-209ggm,intermediates-zcf02673,ksc497i9n,rarechem al be 1247
2734840
3-chloro-5-fluorobenzoic acid
C1=C(C=C(C=C1F)Cl)C(=O)O
25026-64-6
C7H4ClFO2
174.56
MFCD00973904
QFMRANWPGGSNHS-UHFFFAOYSA-N
3-chloro-5-fluoro-benzoic acid,3-fluoro-5-chlorobenzoic acid,5-chloro-3-fluorobenzoic acid,benzoic acid, 3-chloro-5-fluoro,3-chloro-5-fluorobenzoicacid,pubchem1373,acmc-209ggm,intermediates-zcf02673,ksc497i9n,rarechem al be 1247
2734840
3-chloro-5-fluorobenzoic acid
C1=C(C=C(C=C1F)Cl)C(=O)O
Melting Point 153°C to 154°C
Infrared Spectrum Authentic
Quantity 1g
Packaging Glass bottle
Color White to Beige
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (GC)
Linear Formula FC6H3(Cl)COOH
Formula Weight 174.56
Physical Form Crystalline Powder and Granules
Percent Purity 97%
Chemical Name or Material 3-Chloro-5-fluorobenzoic acid
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning