Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Chloro-5-fluorobenzonitrile, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
327056-73-5
C7H3ClFN
155.56
MFCD03407967
GIKLRWNRBOLRDV-UHFFFAOYSA-N
3-chloro-5-fluoro benzonitrile,3-chloro-5-fluoro-benzonitrile,benzonitrile, 3-chloro-5-fluoro,5-chloro-3-fluorobenzenecarbonitrile,5-chloro-3-fluorobenzonitrile,pubchem4725,acmc-1afkx,3-fluoro-5-chloro-benzonitrile,5-fluoro-3-chloro-benzonitrile,buttpark 45\01-07
2734841
3-chloro-5-fluorobenzonitrile
C1=C(C=C(C=C1F)Cl)C#N
327056-73-5
C7H3ClFN
155.56
MFCD03407967
GIKLRWNRBOLRDV-UHFFFAOYSA-N
3-chloro-5-fluoro benzonitrile,3-chloro-5-fluoro-benzonitrile,benzonitrile, 3-chloro-5-fluoro,5-chloro-3-fluorobenzenecarbonitrile,5-chloro-3-fluorobenzonitrile,pubchem4725,acmc-1afkx,3-fluoro-5-chloro-benzonitrile,5-fluoro-3-chloro-benzonitrile,buttpark 45\01-07
2734841
3-chloro-5-fluorobenzonitrile
C1=C(C=C(C=C1F)Cl)C#N
Melting Point 53.0°C to 57.0°C
Infrared Spectrum Authentic
Packaging Glass bottle
Color Yellow
Quantity 5g
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (GC)
Linear Formula ClC6H3(F)CN
Formula Weight 155.56
Physical Form Crystalline Powder
Percent Purity 98%
Chemical Name or Material 3-Chloro-5-fluorobenzonitrile
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Harmful in contact with skin.
May cause respiratory irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning