Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-chloro-5-fluorobenzyl Bromide, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
493024-39-8
C7H5BrClF
223.47
MFCD03788472
PKSAHOPFPPAUOD-UHFFFAOYSA-N
3-chloro-5-fluorobenzyl bromide,1-bromomethyl-3-chloro-5-fluorobenzene,3-chloro-5-fluorobenzylbromide,5-chloro-3-fluorobenzyl bromide,benzene, 1-bromomethyl-3-chloro-5-fluoro,5-bromomethyl-3-chloro-1-fluorobenzene,alpha-bromo-3-chloro-5-fluorotoluene,pubchem4896,acmc-209kfr,1-bromomethyl-3-chloro-5-fluoro-benzene
2734834
1-(bromomethyl)-3-chloro-5-fluorobenzene
C1=C(C=C(C=C1F)Cl)CBr
493024-39-8
C7H5BrClF
223.47
MFCD03788472
PKSAHOPFPPAUOD-UHFFFAOYSA-N
3-chloro-5-fluorobenzyl bromide,1-bromomethyl-3-chloro-5-fluorobenzene,3-chloro-5-fluorobenzylbromide,5-chloro-3-fluorobenzyl bromide,benzene, 1-bromomethyl-3-chloro-5-fluoro,5-bromomethyl-3-chloro-1-fluorobenzene,alpha-bromo-3-chloro-5-fluorotoluene,pubchem4896,acmc-209kfr,1-bromomethyl-3-chloro-5-fluoro-benzene
2734834
1-(bromomethyl)-3-chloro-5-fluorobenzene
C1=C(C=C(C=C1F)Cl)CBr
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 5g
Color White
Melting Point 25°C to 30°C
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (GC)
Linear Formula ClC6H3(F)CH2Br
Formula Weight 223.47
Physical Form Low Melting Crystals, Crystalline Mass or Liquid After Melting
Percent Purity 97%
Chemical Name or Material 3-Chloro-5-fluorobenzyl bromide
Show More Show Less

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger