Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ 3-Chloro-5-fluoropyridine, 98%
Click to view available options
Quantity:
1g
5g
Specifications
Flash Point | 41°C (106°F) |
Refractive Index | 1.5015 |
Melting Point | 25°C to 27°C |
Quantity | 1g |
UN Number | UN1325 |
Formula Weight | 131.54 |
Percent Purity | 98% |
Chemical Name or Material | 3-Chloro-5-fluoropyridine |
Safety and Handling
GHS H Statement
H318-H228-H302-H312-H332-H335-H315
Causes serious eye damage.
Flammable solid.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
May cause respiratory irritation.
Causes skin irritation.
P210-P240-P241-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P370+P378q-P501c
H228-H302+H312+H332-H315-H318-H335
DOTInformation : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLIDS, ORGANIC, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures