Alfa Aesar™ 3-Chloro-5-(trifluoromethyl)benzeneboronic acid pinacol ester, 97%

Manufacturer: Alfa Aesar™ H5171203

Quantity 1g

 View more versions of this product

Catalog No. AAH5171203

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 942069-65-0
    Molecular Formula C13H15BClF3O2
    Formula Weight 306.52
    MDL Number MFCD11856039
    Synonym 2-[3-Chloro-5-(trifluoromethyl)phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
    Chemical Name or Material 3-Chloro-5-(trifluoromethyl)benzeneboronic acid pinacol ester

    Specifications

    Formula Weight 306.52
    Molecular Formula C13H15BClF3O2
    MDL Number MFCD11856039
    CAS 942069-65-0
    Synonym 2-[3-Chloro-5-(trifluoromethyl)phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
    Quantity 1g
    Chemical Name or Material 3-Chloro-5-(trifluoromethyl)benzeneboronic acid pinacol ester
    Assay 97%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit