Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Chloro-6-methoxypyridazine, ACROS Organics™

Click to view available options
Quantity:
5g
25g
1722-10-7
C5H5ClN2O
144.558
XBJLKXOOHLLTPG-UHFFFAOYSA-N
pyridazine, 3-chloro-6-methoxy,3-chloro-6-methoxy-pyridazine,6-chloro-3-methoxypyridazine,pyridazine,3-chloro-6-methoxy,6-chloropyridazin-3-yl methyl ether,3-chloro-6-methoxypyridazine,zlchem 731,acmc-1bvtl,3-methoxy-6-chloropyridazine,3-chloro-6-methoxy pyridazine
74403
3-chloro-6-methoxypyridazine
COC1=NN=C(C=C1)Cl
1722-10-7
C5H5ClN2O
144.558
XBJLKXOOHLLTPG-UHFFFAOYSA-N
pyridazine, 3-chloro-6-methoxy,3-chloro-6-methoxy-pyridazine,6-chloro-3-methoxypyridazine,pyridazine,3-chloro-6-methoxy,6-chloropyridazin-3-yl methyl ether,3-chloro-6-methoxypyridazine,zlchem 731,acmc-1bvtl,3-methoxy-6-chloropyridazine,3-chloro-6-methoxy pyridazine
74403
3-chloro-6-methoxypyridazine
COC1=NN=C(C=C1)Cl
Quantity 5g
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Formula Weight 144.56
Chemical Name or Material 3-Chloro-6-methoxypyridazine, 97%