Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
3-Chloro-alpha-toluenesulfonyl chloride, 96%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
Description
3-Chloro-alpha-toluenesulfonyl chloride is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3-Chloro-alpha-toluenesulfonyl chloride is used as a pharmaceutical intermediate.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Store under dry inert gas. It is hygroscopic in nature. Incompatible with oxidizing agents. It is sensitive to moisture.
3-Chloro-alpha-toluenesulfonyl chloride is used as a pharmaceutical intermediate.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Store under dry inert gas. It is hygroscopic in nature. Incompatible with oxidizing agents. It is sensitive to moisture.
Specifications
Melting Point | 69°C to 75°C |
Quantity | 1 g |
UN Number | UN3261 |
Formula Weight | 225.09 |
Percent Purity | 96% |
Chemical Name or Material | 3-Chloro-alpha-toluenesulfonyl chloride |
Safety and Handling
Hazard Category | H302+H312+H332-H314-H335 |
Hazard Statement | GHS H Statement H314-H318 Causes severe skin burns and eye damage. Causes serious eye damage. |
Precautionary Statement | P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c |
DOTInformation | Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S. |
TSCA | No |
Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction