Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Chloro-N-Fmoc-L-phenylalanine, 95%

Click to view available options
Quantity:
1g
5g
198560-44-0
C24H20ClNO4
421.877
MFCD00672548
UOZAKKJRIKXQPY-QFIPXVFZSA-N
fmoc-l-3-chlorophenylalanine,fmoc-3-chloro-l-phenylalanine,fmoc-phe 3-cl-oh,fmoc-l-phe 3-cl-oh,l-3-3-chlorophenyl-n-fmoc-alanine,fmoc-l-3-chlorophe,l-3-chlorophenyl-n-fmoc-alanine,s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-3-chlorophenyl propanoic acid,2s-3-3-chlorophenyl-2-9h-fluoren-9-ylmethoxycarbonylamino propanoic acid,2s-3-3-chlorophenyl-2-9h-fluoren-9-ylmethoxy carbonyl amino propanoic acid
2761466
(2S)-3-(3-chlorophenyl)-2-(9H-fluoren-9-ylmethoxycarbonylamino)propanoic acid
C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CC4=CC(=CC=C4)Cl)C(=O)O
198560-44-0
C24H20ClNO4
421.877
MFCD00672548
UOZAKKJRIKXQPY-QFIPXVFZSA-N
fmoc-l-3-chlorophenylalanine,fmoc-3-chloro-l-phenylalanine,fmoc-phe 3-cl-oh,fmoc-l-phe 3-cl-oh,l-3-3-chlorophenyl-n-fmoc-alanine,fmoc-l-3-chlorophe,l-3-chlorophenyl-n-fmoc-alanine,s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-3-chlorophenyl propanoic acid,2s-3-3-chlorophenyl-2-9h-fluoren-9-ylmethoxycarbonylamino propanoic acid,2s-3-3-chlorophenyl-2-9h-fluoren-9-ylmethoxy carbonyl amino propanoic acid
2761466
(2S)-3-(3-chlorophenyl)-2-(9H-fluoren-9-ylmethoxycarbonylamino)propanoic acid
C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CC4=CC(=CC=C4)Cl)C(=O)O
Formula Weight 421.88
Physical Form Solid
Percent Purity 95%
Quantity 1g
Chemical Name or Material 3-Chloro-N-Fmoc-L-phenylalanine

storageNote1 : 2° to 8°C