Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Chloro-N-methylbenzylamine, 97%

Manufacturer:  Alfa Aesar™ H5498603

Quantity 1g

 View more versions of this product

Catalog No. AAH5498603


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

39191-07-6
155.63
UN1993
C8H10ClN
MFCD00045194
3-Chloro-N-methylbenzylamine

Specifications

46°C (115°F)
1g
C8H10ClN
MFCD00045194
3-Chloro-N-methylbenzylamine
97°C
39191-07-6
155.63
UN1993
97%

Safety and Handling

SDS & Safety Documents

GHS H Statement
H318-H226-H302-H315
Causes serious eye damage.
Flammable liquid and vapor.
Harmful if swallowed.
Causes skin irritation.

GHS P Statement
P210-P280-P303+P361+P353-P305+P351+P338-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit