Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3'-Chloroacetophenone, 98+%

Click to view available options
Quantity:
5g
25g
100g
99-02-5
C8H7ClO
154.593
MFCD00000593
UUWJBXKHMMQDED-UHFFFAOYSA-N
3'-chloroacetophenone,1-3-chlorophenyl ethanone,m-chloroacetophenone,ethanone, 1-3-chlorophenyl,3-chloroacetophenone,1-3-chlorophenyl ethan-1-one,3-chloro-acetophenone,acetophenone, 3'-chloro,1-3-chloro-phenyl-ethanone,3/'-chloroacetophenone
14933
1-(3-chlorophenyl)ethanone
CC(=O)C1=CC(=CC=C1)Cl
99-02-5
C8H7ClO
154.593
MFCD00000593
UUWJBXKHMMQDED-UHFFFAOYSA-N
3'-chloroacetophenone,1-3-chlorophenyl ethanone,m-chloroacetophenone,ethanone, 1-3-chlorophenyl,3-chloroacetophenone,1-3-chlorophenyl ethan-1-one,3-chloro-acetophenone,acetophenone, 3'-chloro,1-3-chloro-phenyl-ethanone,3/'-chloroacetophenone
14933
1-(3-chlorophenyl)ethanone
CC(=O)C1=CC(=CC=C1)Cl
Quantity 100g
Refractive Index 1.5506
Density 1.191
Flash Point 105°C (221°F)
Boiling Point 227°C to 229°C
UN Number UN3416
Beilstein 636318
Formula Weight 154.6
Percent Purity ≥98%
Chemical Name or Material 3'-Chloroacetophenone

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P311-P330-P332+P313-P362-P501c

H302-H315-H319-H331-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: CHLOROACETOPHENONE, LIQUID

EINECSNumber : 202-721-8

TSCA : Yes

Recommended Storage : Ambient temperatures