Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3'-Chloroacetophenone 98%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
Packaging:
Glass bottle
99-02-5
C8H7ClO
154.6
MFCD00000593
UUWJBXKHMMQDED-UHFFFAOYSA-N
3'-chloroacetophenone,1-3-chlorophenyl ethanone,m-chloroacetophenone,ethanone, 1-3-chlorophenyl,3-chloroacetophenone,1-3-chlorophenyl ethan-1-one,3-chloro-acetophenone,acetophenone, 3'-chloro,1-3-chloro-phenyl-ethanone,3/'-chloroacetophenone
14933
1-(3-chlorophenyl)ethanone
CC(=O)C1=CC(=CC=C1)Cl
99-02-5
C8H7ClO
154.6
MFCD00000593
UUWJBXKHMMQDED-UHFFFAOYSA-N
3'-chloroacetophenone,1-3-chlorophenyl ethanone,m-chloroacetophenone,ethanone, 1-3-chlorophenyl,3-chloroacetophenone,1-3-chlorophenyl ethan-1-one,3-chloro-acetophenone,acetophenone, 3'-chloro,1-3-chloro-phenyl-ethanone,3/'-chloroacetophenone
14933
1-(3-chlorophenyl)ethanone
CC(=O)C1=CC(=CC=C1)Cl
Quantity 100g
Refractive Index 1.5480 to 1.55
Density 1.1900g/mL
Specific Gravity 1.19
Boiling Point 227°C to 229°C
Flash Point 105°C
Packaging Glass bottle
Infrared Spectrum Authentic
Color Colorless to Yellow
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Beilstein 07, 282
Merck Index 10, 2082
Formula Weight 154.6
Physical Form Liquid
Percent Purity ≥97.5%
Chemical Name or Material 3'-Chloroacetophenone, 98%
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Harmful if swallowed.
Causes skin irritation.
Fatal if inhaled.
May cause respiratory irritation.

GHS P Statement:
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 202-721-8

TSCA : TSCA