Alfa Aesar™ 3-Chloroaniline, 99%

Manufacturer: Alfa Aesar™ A1201922

Quantity 100g

 View more versions of this product

Catalog No. AAA1201922

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 108-42-9
    Molecular Formula C6H6ClN
    Formula Weight 127.57
    MDL Number MFCD00007765
    UN Number UN2019
    Beilstein 605969
    Merck Index 14,2118
    Chemical Name or Material 3-Chloroaniline

    Specifications

    Formula Weight 127.57
    Health Hazard 3 GHS P Statement
    P280h-P273-P309-P310-P302+P352-P501a
    Wear protective gloves/clothing.
    Avoid release to the environment.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN: Wash with plenty of soap and water.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    MDL Number MFCD00007765
    RTECS Number BX0350000
    CAS 108-42-9
    Health Hazard 1 Danger
    Odor Characteristic
    Boiling Point 230°C
    Molecular Formula C6H6ClN
    UN Number UN2019
    Beilstein 605969
    Merck Index 14,2118
    Health Hazard 2 GHS H Statement
    H301-H311-H331-H373
    Toxic if swallowed.
    Toxic in contact with skin.
    Toxic if inhaled.
    May cause damage to organs through prolonged or repeated exposure.
    DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: CHLOROANILINES, LIQUID
    Flash Point 123°C (253°F)
    Melting Point −10°C
    Quantity 100g
    TSCA TSCA
    Chemical Name or Material 3-Chloroaniline
    Assay 99%

    Description

    Applications
    3-Chloroaniline was the only aniline compound to increase plasmaALT/GPT activity at 48 h. It is employed in the bioaugmentation of activated sludge.

    Solubility
    Soluble in water. [6.8 g/L (20°C)].

    Notes
    Store in cool place. Keep away from oxidizing agents. Incompatible with bases.

    GHS H Statement
    H301-H311-H331-H373
    Toxic if swallowed.
    Toxic in contact with skin.
    Toxic if inhaled.
    May cause damage to organs through prolonged or repeated exposure.

    GHS P Statement
    P280h-P273-P309-P310-P302+P352-P501a
    Wear protective gloves/clothing.
    Avoid release to the environment.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN: Wash with plenty of soap and water.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit