Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Chlorobenzamide, 98%, ACROS Organics™

Click to view available options
Quantity:
25g
Packaging:
Glass bottle
618-48-4
C7H6ClNO
155.581
MFCD00014799
MJTGQALMWUUPQM-UHFFFAOYSA-N
m-chlorobenzamide,benzamide, 3-chloro,3-chloro-benzamide,benzamide, m-chloro-8ci,benzamide, m-chloro,5-chlorobenzamide,pubchem3618,acmc-1b3cu,wce
69254
CHEBI:10587
3-chlorobenzamide
C1=CC(=CC(=C1)Cl)C(=O)N
618-48-4
C7H6ClNO
155.581
MFCD00014799
MJTGQALMWUUPQM-UHFFFAOYSA-N
m-chlorobenzamide,benzamide, 3-chloro,3-chloro-benzamide,benzamide, m-chloro-8ci,benzamide, m-chloro,5-chlorobenzamide,pubchem3618,acmc-1b3cu,wce
69254
CHEBI:10587
3-chlorobenzamide
C1=CC(=CC(=C1)Cl)C(=O)N
Infrared Spectrum Authentic
Color White
Melting Point 133°C to 137°C
Packaging Glass bottle
Quantity 25g
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula ClC6H4C(O)NH2
Formula Weight 155.58
Physical Form Powder
Percent Purity ≥97.5%
Chemical Name or Material 3-Chlorobenzamide, 98%
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Harmful if inhaled.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.