Learn More
3-Chlorobenzeneboronic acid, 97%, Thermo Scientific Chemicals
Description
It is employed as a reactant involved in 1,4-conjugate addition reactions with ethenesulfonamides to form arylethanesulfonamides, Suzuki-Miyaura reactions with dibromotrifluoromethylbenzene, cross-coupling reactions with diazoesters3 or potassium cyanate. It is involved in the synthesis of biarylketones and phthalides and synthesis of inhibitors including PDE4 inhibitors6 among others.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
It is employed as a reactant involved in 1,4-conjugate addition reactions with ethenesulfonamides to form arylethanesulfonamides, Suzuki-Miyaura reactions with dibromotrifluoromethylbenzene, cross-coupling reactions with diazoesters3 or potassium cyanate. It is involved in the synthesis of biarylketones and phthalides and synthesis of inhibitors including PDE4 inhibitors6 among others.
Solubility
Slightly soluble in water. soluble in ether, tetrahydrofuran,, dimethyl sulfoxide, dimethyl formamide, methanol.
Notes
Store at room temperature. Incompatible with oxidizing agents.
Specifications
Melting Point | 180°C to 185°C |
Quantity | 5 g |
Beilstein | 2936343 |
Solubility Information | Slightly soluble in water. soluble in ether,tetrahydrofuran,,dimethyl sulfoxide,dimethyl formamide,methanol. |
Formula Weight | 156.38 |
Percent Purity | 97% |
Chemical Name or Material | 3-Chlorobenzeneboronic acid |
Safety and Handling
Hazard Category | H302-H312-H315-H319-H332-H335 |
Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
Precautionary Statement | P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c |
EINECSNumber | 000-000-0 |
TSCA | No |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.