Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Chlorobenzo[b]thiophene-2-carbonyl chloride, 97%, Maybridge™

Click to view available options
Quantity:
1g
10g
25g
21815-91-8
C9H4Cl2OS
231.09
MFCD00053069
GWKSSMDJEWPKCM-UHFFFAOYSA-N
3-chlorobenzo b thiophene-2-carbonyl chloride, 3-chlorobenzothiophene-2-carbonyl chloride, 3-chloro-benzo b thiophene-2-carbonyl chloride, 3-chlorobenzo b thiophen-2-carbonyl chloride, benzo b thiophene-2-carbonyl chloride, 3-chloro, 3-chlorobenzo b-2-thiophenecarboxylic acid chloride, acmc-1cne6, 3-chlorobenzo b thiophene-2-carbonylchloride, 2-chlorocarbonyl-3-chlorobenzo b thiophene, 3-chlorobenzo thiophene-2-carbonyl chloride
519898
3-chloro-1-benzothiophene-2-carbonyl chloride
ClC(=O)C1=C(Cl)C2=CC=CC=C2S1
21815-91-8
C9H4Cl2OS
231.09
MFCD00053069
GWKSSMDJEWPKCM-UHFFFAOYSA-N
3-chlorobenzo b thiophene-2-carbonyl chloride, 3-chlorobenzothiophene-2-carbonyl chloride, 3-chloro-benzo b thiophene-2-carbonyl chloride, 3-chlorobenzo b thiophen-2-carbonyl chloride, benzo b thiophene-2-carbonyl chloride, 3-chloro, 3-chlorobenzo b-2-thiophenecarboxylic acid chloride, acmc-1cne6, 3-chlorobenzo b thiophene-2-carbonylchloride, 2-chlorocarbonyl-3-chlorobenzo b thiophene, 3-chlorobenzo thiophene-2-carbonyl chloride
519898
3-chloro-1-benzothiophene-2-carbonyl chloride
ClC(=O)C1=C(Cl)C2=CC=CC=C2S1
Quantity 25g
Packaging Amber Glass Bottle
Assay Percent Range 97%
Formula Weight 231.1
Chemical Name or Material 3-Chlorobenzo[b]thiophene-2-carbonyl chloride