Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Chlorobenzophenone, 98+%

Click to view available options
Quantity:
5g
25g
100g
1016-78-0
C13H9ClO
216.664
MFCD00009816
CPLWKNRPZVNELG-UHFFFAOYSA-N
3-chlorobenzophenone,3-chlorophenyl phenyl methanone,benzophenone, 3-chloro,methanone, 3-chlorophenyl phenyl,m-chlorobenzophenone,3-chlorophenyl-phenylmethanone,methanone, 3-chlorophenyl pheny,3-chlorophenyl phenyl ketone,5-chlorobenzophenone,pubchem24414
66098
(3-chlorophenyl)-phenylmethanone
C1=CC=C(C=C1)C(=O)C2=CC(=CC=C2)Cl
1016-78-0
C13H9ClO
216.664
MFCD00009816
CPLWKNRPZVNELG-UHFFFAOYSA-N
3-chlorobenzophenone,3-chlorophenyl phenyl methanone,benzophenone, 3-chloro,methanone, 3-chlorophenyl phenyl,m-chlorobenzophenone,3-chlorophenyl-phenylmethanone,methanone, 3-chlorophenyl pheny,3-chlorophenyl phenyl ketone,5-chlorobenzophenone,pubchem24414
66098
(3-chlorophenyl)-phenylmethanone
C1=CC=C(C=C1)C(=O)C2=CC(=CC=C2)Cl
Quantity 100g
Melting Point 81°C to 85°C
Beilstein 1869822
Formula Weight 216.67
Percent Purity ≥98%
Chemical Name or Material 3-Chlorobenzophenone