Alfa Aesar™ 3-Chlorobenzyl alcohol, 99%

Manufacturer: Alfa Aesar™ A1184118

Quantity 50g

 View more versions of this product

Catalog No. AAA1184118

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 873-63-2
    Molecular Formula C7H7ClO
    Formula Weight 142.59
    MDL Number MFCD00004632
    Beilstein 2041501
    Chemical Name or Material 3-Chlorobenzyl alcohol

    Specifications

    Formula Weight 142.59
    Molecular Formula C7H7ClO
    CAS 873-63-2
    Flash Point >110°C (230°F)
    Boiling Point 236° to 237°C
    MDL Number MFCD00004632
    Quantity 50g
    Beilstein 2041501
    Chemical Name or Material 3-Chlorobenzyl alcohol
    Assay 99%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit