Alfa Aesar™ 3-Chlorobenzyl chloride, 97%

Manufacturer: Alfa Aesar™ A1104114

Quantity 25g

 View more versions of this product

Catalog No. AAA1104114

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 620-20-2
    Molecular Formula C7H6Cl2
    Formula Weight 161.03
    MDL Number MFCD00000905
    UN Number UN2235
    Beilstein 742266
    Synonym alpha,3-Dichlorotoluene
    Chemical Name or Material 3-Chlorobenzyl chloride

    Specifications

    Formula Weight 161.03
    Health Hazard 3 GHS P Statement
    P280-P273-P305+P351+P338-P309-P310
    Wear protective gloves/protective clothing/eye protection/face protection.
    Avoid release to the environment.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
    Beilstein 742266
    Synonym alpha,3-Dichlorotoluene
    Health Hazard 2 GHS H Statement
    H314-H318-H302-H312-H332
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Harmful if swallowed.
    Harmful in contact with skin.
    Harmful if inhaled.
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: CHLOROBENZYL CHLORIDES, LIQUID
    Flash Point 98°C (208°F)
    Boiling Point 215° to 216°C
    TSCA TSCA
    Odor Pungent
    Molecular Formula C7H6Cl2
    MDL Number MFCD00000905
    CAS 620-20-2
    UN Number UN2235
    Health Hazard 1 Danger
    Quantity 25g
    Chemical Name or Material 3-Chlorobenzyl chloride
    Assay 97%

    GHS H Statement
    H314-H318-H302-H312-H332
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Harmful if swallowed.
    Harmful in contact with skin.
    Harmful if inhaled.

    GHS P Statement
    P280-P273-P305+P351+P338-P309-P310
    Wear protective gloves/protective clothing/eye protection/face protection.
    Avoid release to the environment.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit