Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Chlorobenzyl Chloride, 98+%, ACROS Organics™

Click to view available options
Quantity:
25mL
Packaging:
Glass bottle
620-20-2
C7H6Cl2
161.025
MFCD00000905
DDGRAFHHXYIQQR-UHFFFAOYSA-N
3-chlorobenzyl chloride,1-chloro-3-chloromethyl benzene,alpha,3-dichlorotoluene,m-chlorobenzyl chloride,benzene, 1-chloro-3-chloromethyl,3-chlorobenzylchloride,m-chlorbenzyl chloride,1-chloro-3-chloromethyl-benzene,toluene, m,.alpha.-dichloro,m,.alpha.-dichlorotoluene
12103
1-chloro-3-(chloromethyl)benzene
C1=CC(=CC(=C1)Cl)CCl
620-20-2
C7H6Cl2
161.025
MFCD00000905
DDGRAFHHXYIQQR-UHFFFAOYSA-N
3-chlorobenzyl chloride,1-chloro-3-chloromethyl benzene,alpha,3-dichlorotoluene,m-chlorobenzyl chloride,benzene, 1-chloro-3-chloromethyl,3-chlorobenzylchloride,m-chlorbenzyl chloride,1-chloro-3-chloromethyl-benzene,toluene, m,.alpha.-dichloro,m,.alpha.-dichlorotoluene
12103
1-chloro-3-(chloromethyl)benzene
C1=CC(=CC(=C1)Cl)CCl
Quantity 25mL
Packaging Glass bottle
Color Colorless to Yellow
Infrared Spectrum Authentic
Refractive Index 1.5540 to 1.5560 (20°C, 589nm)
CAS Min % 98.0
CAS Max % 100.0
Assay Percent Range 98+%
Linear Formula ClC6H4CH2Cl
Beilstein 05, II, 231
Formula Weight 161.03
Physical Form Liquid
Percent Purity ≥98.0%
Chemical Name or Material 3-Chlorobenzyl chloride
Show More Show Less

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful if inhaled.
Harmful in contact with skin.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger