Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
3-Chlorobenzylmagnesium chloride, 0.50M in 2-MeTHF, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 mL
Description
3-Chlorobenzylmagnesium chloride is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3-Chlorobenzylmagnesium chloride is used as pharmaceutical intermediate.
Solubility
It reacts with water.
Notes
Air and moisture sensitive. Incompatible with oxidizing agents and acids.
3-Chlorobenzylmagnesium chloride is used as pharmaceutical intermediate.
Solubility
It reacts with water.
Notes
Air and moisture sensitive. Incompatible with oxidizing agents and acids.
Specifications
Quantity | 100 mL |
UN Number | UN2924 |
Solubility Information | It reacts with water. |
Formula Weight | 185.33 |
Concentration or Composition (by Analyte or Components) | 0.50M in 2-MeTHF |
Chemical Name or Material | 3-Chlorobenzylmagnesium chloride |
Safety and Handling
Hazard Category | H225-H302-H314-H335 |
Hazard Statement | GHS H Statement H225-H314 Highly flammable liquid and vapor. Causes severe skin burns and eye damage. |
Precautionary Statement | P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c |
DOTInformation | Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S. |
TSCA | No |
Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction