Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Chlorobenzylsulfonyl chloride, 97%, Acros Organics™

Click to view available options
Quantity:
1g
5g
25g
24974-73-0
C7H6Cl2O2S
225.09
LXTGNVLBPVVMSL-UHFFFAOYSA-N
3-chlorophenyl methanesulfonyl chloride,3-chloro-phenyl-methanesulfonyl chloride,chloro 3-chlorophenyl methyl sulfone,benzenemethanesulfonyl chloride, 3-chloro,pubchem5501,acmc-20an5u,3-chlorobenzylsulfonyl chloride,3-chlorophenyl methanesulfonylchloride,benzenemethanesulfonylchloride,3-chloro,benzenemethanesulfonylchloride, 3-chloro
2757802
(3-chlorophenyl)methanesulfonyl chloride
C1=CC(=CC(=C1)Cl)CS(=O)(=O)Cl
24974-73-0
C7H6Cl2O2S
225.09
LXTGNVLBPVVMSL-UHFFFAOYSA-N
3-chlorophenyl methanesulfonyl chloride,3-chloro-phenyl-methanesulfonyl chloride,chloro 3-chlorophenyl methyl sulfone,benzenemethanesulfonyl chloride, 3-chloro,pubchem5501,acmc-20an5u,3-chlorobenzylsulfonyl chloride,3-chlorophenyl methanesulfonylchloride,benzenemethanesulfonylchloride,3-chloro,benzenemethanesulfonylchloride, 3-chloro
2757802
(3-chlorophenyl)methanesulfonyl chloride
C1=CC(=CC(=C1)Cl)CS(=O)(=O)Cl
Quantity 5g
Melting Point 71.0°C to 75.0°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. As diethylamine derivative (HPLC)
Formula Weight 225.09
Percent Purity 97%
Chemical Name or Material 3-Chlorobenzylsulfonyl chloride

GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Take off im

GHS Signal Word: Danger