Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3'-Chlorobiphenyl-4-sulfonyl chloride, 95%, Acros Organics

Click to view available options
Quantity:
1g
5g
478647-00-6
C12H8Cl2O2S
287.17
FVELIXWAKJVRDG-UHFFFAOYSA-N
3'-chloro-1,1'-biphenyl-4-sulfonyl chloride,3'-chloro-biphenyl-4-sulfonyl chloride,3'-chloro-biphenyl-4-sulfonylchloride,3'-chlorobiphenyl-4-sulfonyl chloride,bis-o,o-diethylphosphorothionicanhydrid,3'-chloro-1,1'-biphenyl-4-sulfonylchloride,3'-chloro 1,1'-biphenyl-4-sulfonyl chloride,1,1'-biphenyl-4-sulfonylchloride,3'-chloro,4-3-chlorophenyl benzenesulfonyl chloride
16244688
4-(3-chlorophenyl)benzenesulfonyl chloride
C1=CC(=CC(=C1)Cl)C2=CC=C(C=C2)S(=O)(=O)Cl
478647-00-6
C12H8Cl2O2S
287.17
FVELIXWAKJVRDG-UHFFFAOYSA-N
3'-chloro-1,1'-biphenyl-4-sulfonyl chloride,3'-chloro-biphenyl-4-sulfonyl chloride,3'-chloro-biphenyl-4-sulfonylchloride,3'-chlorobiphenyl-4-sulfonyl chloride,bis-o,o-diethylphosphorothionicanhydrid,3'-chloro-1,1'-biphenyl-4-sulfonylchloride,3'-chloro 1,1'-biphenyl-4-sulfonyl chloride,1,1'-biphenyl-4-sulfonylchloride,3'-chloro,4-3-chlorophenyl benzenesulfonyl chloride
16244688
4-(3-chlorophenyl)benzenesulfonyl chloride
C1=CC(=CC(=C1)Cl)C2=CC=C(C=C2)S(=O)(=O)Cl
Melting Point 71°C to 74°C
Quantity 5g
Assay Percent Range 94% min. Diethylsulfonamide derivative (HPLC)
Formula Weight 287.17
Percent Purity 95%
Chemical Name or Material 3'-Chlorobiphenyl-4-sulfonyl chloride

GHS H Statement
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Take off im

GHS Signal Word: Danger