Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
3-Chloroheptafluoro-2-butene, 96%
Click to view available options
Quantity:
5g
Specifications
Quantity | 5g |
Boiling Point | 31°C to 32°C |
Refractive Index | 1.294 |
Density | 1.548 |
Beilstein | 1783461 |
Formula Weight | 216.49 |
Percent Purity | 96% |
Chemical Name or Material | 3-Chloroheptafluoro-2-butene |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 000-000-0
TSCA : No
Recommended Storage : Keep cold