Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Chloromandelic acid, 97%

Click to view available options
Quantity:
5g
25g
100g
16273-37-3
C8H7ClO3
186.591
MFCD00829409
SAMVPMGKGGLIPF-UHFFFAOYSA-N
3-chloromandelic acid,2-3-chlorophenyl-2-hydroxyacetic acid,3-chlorophenylglycolic acid,3-chlorophenyl hydroxy acetic acid,3-chloromandelicacid,benzeneacetic acid,3-chloro-a-hydroxy-, ar,maybridge3_000705,3-chloro-dl-mandelic acid,acmc-1c3h4,racemic 3-chloromandelic acid
85981
2-(3-chlorophenyl)-2-hydroxyacetic acid
C1=CC(=CC(=C1)Cl)C(C(=O)O)O
16273-37-3
C8H7ClO3
186.591
MFCD00829409
SAMVPMGKGGLIPF-UHFFFAOYSA-N
3-chloromandelic acid,2-3-chlorophenyl-2-hydroxyacetic acid,3-chlorophenylglycolic acid,3-chlorophenyl hydroxy acetic acid,3-chloromandelicacid,benzeneacetic acid,3-chloro-a-hydroxy-, ar,maybridge3_000705,3-chloro-dl-mandelic acid,acmc-1c3h4,racemic 3-chloromandelic acid
85981
2-(3-chlorophenyl)-2-hydroxyacetic acid
C1=CC(=CC(=C1)Cl)C(C(=O)O)O
Quantity 100g
Formula Weight 186.6
Percent Purity 97%
Chemical Name or Material 3-Chloromandelic acid

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning