Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(chloromethyl)heptane, 90%, ACROS Organics™

Click to view available options
Quantity:
25mL
100mL
Packaging:
Glass Bottle
123-04-6
C8H17Cl
148.674
MFCD00018940
WLVCBAMXYMWGLJ-UHFFFAOYSA-N
3-chloromethyl heptane,2-ethylhexyl chloride,1-chloro-2-ethylhexane,heptane, 3-chloromethyl,hexane, 1-chloro-2-ethyl,2-ethylhexylchlorid,2-ethylhexylchloride,acmc-209any,4-01-00-00430 beilstein handbook reference
31240
3-(chloromethyl)heptane
CCCCC(CC)CCl
123-04-6
C8H17Cl
148.674
MFCD00018940
WLVCBAMXYMWGLJ-UHFFFAOYSA-N
3-chloromethyl heptane,2-ethylhexyl chloride,1-chloro-2-ethylhexane,heptane, 3-chloromethyl,hexane, 1-chloro-2-ethyl,2-ethylhexylchlorid,2-ethylhexylchloride,acmc-209any,4-01-00-00430 beilstein handbook reference
31240
3-(chloromethyl)heptane
CCCCC(CC)CCl
Packaging Glass Bottle
Flash Point 60°C
Color Yellow
Density 0.8820g/mL
Melting Point -70.0°C
Quantity 25mL
Infrared Spectrum Authentic
Boiling Point 166.0°C to 168.0°C
Refractive Index 1.4320 to 1.4360
CAS Min % 88.0
CAS Max % 100.0
Assay Percent Range 90%
Linear Formula H3C(CH2)3CH(CH2Cl)CH2CH3
Formula Weight 148.68
Physical Form Liquid
Percent Purity 90%
Chemical Name or Material 3-(Chloromethyl)heptane
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Flammable liquid and vapor.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning