Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Chlorophenethylalcohol, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
Glass bottle,Glass Bottle
5182-44-5
C8H9ClO
156.61
MFCD00002892
NDWAVJKRSASRPH-UHFFFAOYSA-N
2-3-chlorophenyl ethanol,3-chlorophenethyl alcohol,benzeneethanol, 3-chloro,3-chlorophenethylalcohol,2-3-chlorophenyl ethan-1-ol,m-chlorophenethyl alcohol,m-chlorophenethylic alcohol,phenethyl alcohol, m-chloro,3-chlorobenzeneethanol,acmc-1cuee
78856
2-(3-chlorophenyl)ethanol
C1=CC(=CC(=C1)Cl)CCO
5182-44-5
C8H9ClO
156.61
MFCD00002892
NDWAVJKRSASRPH-UHFFFAOYSA-N
2-3-chlorophenyl ethanol,3-chlorophenethyl alcohol,benzeneethanol, 3-chloro,3-chlorophenethylalcohol,2-3-chlorophenyl ethan-1-ol,m-chlorophenethyl alcohol,m-chlorophenethylic alcohol,phenethyl alcohol, m-chloro,3-chlorobenzeneethanol,acmc-1cuee
78856
2-(3-chlorophenyl)ethanol
C1=CC(=CC(=C1)Cl)CCO
Boiling Point 135.0°C to 137.0°C (13.0mmHg)
Density 1.1810g/mL
Color Colorless to Yellow
Quantity 5g
Infrared Spectrum Authentic
Flash Point >110°C
Specific Gravity 1.181
Packaging Glass bottle
Refractive Index 1.5475 to 1.5495
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (GC)
Linear Formula ClC6H4CH2CH2OH
Formula Weight 156.61
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material 3-Chlorophenethylalcohol, 98%
Show More Show Less

EINECSNumber : 225-964-1

TSCA : TSCA