Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Chlorophenethylalcohol, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass Bottle
5182-44-5
C8H9ClO
156.61
MFCD00002892
NDWAVJKRSASRPH-UHFFFAOYSA-N
2-3-chlorophenyl ethanol, 3-chlorophenethyl alcohol, benzeneethanol, 3-chloro, 3-chlorophenethylalcohol, 2-3-chlorophenyl ethan-1-ol, m-chlorophenethyl alcohol, m-chlorophenethylic alcohol, phenethyl alcohol, m-chloro, 3-chlorobenzeneethanol, acmc-1cuee
78856
2-(3-chlorophenyl)ethan-1-ol
OCCC1=CC=CC(Cl)=C1
5182-44-5
C8H9ClO
156.61
MFCD00002892
NDWAVJKRSASRPH-UHFFFAOYSA-N
2-3-chlorophenyl ethanol, 3-chlorophenethyl alcohol, benzeneethanol, 3-chloro, 3-chlorophenethylalcohol, 2-3-chlorophenyl ethan-1-ol, m-chlorophenethyl alcohol, m-chlorophenethylic alcohol, phenethyl alcohol, m-chloro, 3-chlorobenzeneethanol, acmc-1cuee
78856
2-(3-chlorophenyl)ethan-1-ol
OCCC1=CC=CC(Cl)=C1
Color Colorless to Yellow
Specific Gravity 1.181
Refractive Index 1.5475 to 1.5495
Boiling Point 135.0°C to 137.0°C (13.0mmHg)
Quantity 25g
Infrared Spectrum Authentic
Flash Point >110°C
Density 1.1810g/mL
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula ClC6H4CH2CH2OH
Formula Weight 156.61
Physical Form Liquid
Percent Purity 98%
Packaging Glass Bottle
Chemical Name or Material 3-Chlorophenethylalcohol, 98%
Show More Show Less

EINECSNumber : 225-964-1

TSCA : TSCA