Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Chlorophenylacetone, 94%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass Bottle
14123-60-5
C9H9ClO
168.62
VCNYPJMEQHTAHS-UHFFFAOYSA-N
1-3-chlorophenyl propan-2-one,3-chlorophenylacetone,1-3-chlorophenyl acetone,1-3-chlorophenyl-2-propanone,pubchem19805,3-chlorophenyl-acetone,m-chlorophenyl acetone,2-propanone,1-3-chlorophenyl,1-3-chloro-phenyl-propan-2-one,2-propanone, 1-3-chlorophenyl
2734097
1-(3-chlorophenyl)propan-2-one
CC(=O)CC1=CC(=CC=C1)Cl
14123-60-5
C9H9ClO
168.62
VCNYPJMEQHTAHS-UHFFFAOYSA-N
1-3-chlorophenyl propan-2-one,3-chlorophenylacetone,1-3-chlorophenyl acetone,1-3-chlorophenyl-2-propanone,pubchem19805,3-chlorophenyl-acetone,m-chlorophenyl acetone,2-propanone,1-3-chlorophenyl,1-3-chloro-phenyl-propan-2-one,2-propanone, 1-3-chlorophenyl
2734097
1-(3-chlorophenyl)propan-2-one
CC(=O)CC1=CC(=CC=C1)Cl
Packaging Glass Bottle
Refractive Index 1.5320 to 1.5360
Infrared Spectrum Authentic
Quantity 25g
Color Yellow
Assay Percent Range 92% min. (GC)
Beilstein 07,IV,691
Formula Weight 168.62
Physical Form Liquid
Percent Purity 94%
Chemical Name or Material 3-Chlorophenylacetone
Show More Show Less