Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Chlorophenylhydrazine Sulfate 98.0+%, TCI America™

Click to view available options
Quantity:
5g
25g
41713-37-5
C6H9ClN2O4S
240.658
MFCD00050933
ZQMWFTLJZQAIHV-UHFFFAOYSA-N
20542670
(3-chlorophenyl)hydrazine;sulfuric acid
C1=CC(=CC(=C1)Cl)NN.OS(=O)(=O)O
41713-37-5
C6H9ClN2O4S
240.658
MFCD00050933
ZQMWFTLJZQAIHV-UHFFFAOYSA-N
20542670
(3-chlorophenyl)hydrazine;sulfuric acid
C1=CC(=CC(=C1)Cl)NN.OS(=O)(=O)O
Quantity 25g
Color Undesignated
Formula Weight 383.24
Physical Form Crystal-Powder at 20°C
Percent Purity ≥98.0% (T)
Chemical Name or Material 3-Chlorophenylhydrazine Sulfate

EINECSNumber : (3)-0471

TSCA : No