Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ 3-Chloropropionic acid, 98%
Click to view available options
Quantity:
100g
Specifications
Density | 1.27 |
Boiling Point | 203°C to 205°C |
Quantity | 100g |
Melting Point | 37°C to 41°C |
Flash Point | 122°C (251°F) |
UN Number | UN3261 |
Beilstein | 1098495 |
Merck Index | 14,2159 |
Formula Weight | 108.53 |
Percent Purity | 98% |
Show More |
Safety and Handling
GHS H Statement
H301-H314-H318
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P280-P284-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314
DOTInformation : Hazard Class: 8; Packaging Group: II
EINECSNumber : 203-534-4
RTECSNumber : UE8750000
TSCA : Yes
Recommended Storage : Ambient temperatures