Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Chloropropionic acid, 98%

Click to view available options
Quantity:
100g
500g
107-94-8
C3H5ClO2
108.521
MFCD00002764
QEYMMOKECZBKAC-UHFFFAOYSA-N
3-chloropropionic acid,propanoic acid, 3-chloro,propionic acid, 3-chloro,beta-chloropropionic acid,3-chloropropanic acid,beta-monochloropropionic acid,unii-r5j180fn9z,3-chloro-propionic acid,.beta.-chloropropionic acid,.beta.-monochloropropionic acid
7899
3-chloropropanoic acid
C(CCl)C(=O)O
107-94-8
C3H5ClO2
108.521
MFCD00002764
QEYMMOKECZBKAC-UHFFFAOYSA-N
3-chloropropionic acid,propanoic acid, 3-chloro,propionic acid, 3-chloro,beta-chloropropionic acid,3-chloropropanic acid,beta-monochloropropionic acid,unii-r5j180fn9z,3-chloro-propionic acid,.beta.-chloropropionic acid,.beta.-monochloropropionic acid
7899
3-chloropropanoic acid
C(CCl)C(=O)O
Melting Point 37°C to 41°C
Quantity 500g
Flash Point 122°C (251°F)
Boiling Point 203°C to 205°C
UN Number UN3261
Beilstein 1098495
Merck Index 14,2159
Formula Weight 108.53
Percent Purity 98%
Chemical Name or Material 3-Chloropropionic acid

GHS H Statement
H301-H314-H318
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

RTECSNumber : UE8750000

TSCA : TSCA