Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Chloropropionic acid, 98%, Thermo Scientific™

Catalog No. p-91507
Click to view available options
Quantity:
500 g

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107-94-8
Molecular Formula C3H5ClO2
Molecular Weight (g/mol) 108.52
MDL Number MFCD00002764
InChI Key QEYMMOKECZBKAC-UHFFFAOYSA-N
Synonym 3-chloropropionic acid, propanoic acid, 3-chloro, propionic acid, 3-chloro, beta-chloropropionic acid, 3-chloropropanic acid, beta-monochloropropionic acid, unii-r5j180fn9z, 3-chloro-propionic acid, .beta.-chloropropionic acid, .beta.-monochloropropionic acid
PubChem CID 7899
IUPAC Name 3-chloropropanoic acid
SMILES OC(=O)CCCl

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 37.0°C to 41.0°C
Color White to Beige
Boiling Point 203.0°C to 205.0°C
Flash Point 125°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula ClCH2CH2CO2H
Quantity 500 g
Beilstein 02, 249
Merck Index 15, 2161
Solubility Information Solubility in water: freely soluble. Other solubilities: soluble in alcohol,chloroform and ether,miscible with ether
Formula Weight 108.52
Percent Purity 98%
Physical Form Crystalline Low Melting Solid
Chemical Name or Material 3-Chloropropionic acid, 98%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Corrosive to the respiratory tract.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Do not breat
EINECSNumber 203-534-4
RTECSNumber UE8750000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.