Alfa Aesar™ 3-Chloropropyl acetate, 98%

Manufacturer: Alfa Aesar™ B2501314

Quantity 25g

 View more versions of this product

Catalog No. AAB2501314

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 628-09-1
    Molecular Formula CH3CO2(CH2)3Cl
    Formula Weight 136.58
    MDL Number MFCD00001000
    Synonym Acetic acid 3-chloropropyl ester; 1-Acetoxy-3-chloropropane
    Chemical Name or Material 3-Chloropropyl acetate

    Specifications

    Formula Weight 136.58
    MDL Number MFCD00001000
    Boiling Point 70° to 72°C (15mmHg)
    Molecular Formula CH3CO2(CH2)3Cl
    CAS 628-09-1
    Synonym Acetic acid 3-chloropropyl ester; 1-Acetoxy-3-chloropropane
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H315-H319-H335-H227
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Combustible liquid.
    Health Hazard 3 GHS P Statement
    P210-P261-P280-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    RTECS Number UA8944000
    Flash Point 68°C (154°F)
    Quantity 25g
    Chemical Name or Material 3-Chloropropyl acetate
    Assay 98%

    Description

    Applications
    3-Chloropropyl acetate is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agro chemicals, dyestuff.

    Solubility
    Soluble in water (partly).

    Notes
    Store in a cool, dry place in tightly closed container. Incompatible with oxidizing agents.

    GHS H Statement
    H315-H319-H335-H227
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Combustible liquid.

    GHS P Statement
    P210-P261-P280-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit