Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Chloropropyl Acetate, Acros Organics

Click to view available options
Quantity:
100g
628-09-1
C5H9ClO2
136.575
MFCD00001000
KPOHQIPNNIMWRL-UHFFFAOYSA-N
1-propanol, 3-chloro-, acetate,3-chloropropanol acetate,3-chlorprop-1-ylacetat,gamma-chloropropyl acetate,3-chlorprop-1-ylacetat german,acetic acid 3-chloro-propyl ester,1-acetoxy-3-chloropropane,.gamma.-chloropropyl acetate,3-chloro-propyl acetate,acetic acid 3-chloropropyl ester
12334
3-chloropropyl acetate
CC(=O)OCCCCl
628-09-1
C5H9ClO2
136.575
MFCD00001000
KPOHQIPNNIMWRL-UHFFFAOYSA-N
1-propanol, 3-chloro-, acetate,3-chloropropanol acetate,3-chlorprop-1-ylacetat,gamma-chloropropyl acetate,3-chlorprop-1-ylacetat german,acetic acid 3-chloro-propyl ester,1-acetoxy-3-chloropropane,.gamma.-chloropropyl acetate,3-chloro-propyl acetate,acetic acid 3-chloropropyl ester
12334
3-chloropropyl acetate
CC(=O)OCCCCl
Quantity 100g
Color Colorless
Infrared Spectrum Authentic
Refractive Index 1.4280 to 1.4300 (20°C, 589nm)
CAS Min % 97.5
CAS Max % 100.0
Linear Formula CH3COO(CH2)3Cl
Formula Weight 136.58
Physical Form Liquid
Percent Purity ≥97.5%
Chemical Name or Material 3-Chloropropyl acetate
Show More Show Less