Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ (3-Chloropropyl)trichlorosilane, 97%

Click to view available options
Quantity:
100g
2550-06-3
C12H24O4Si4
344.66
MFCD00013694
VMAWODUEPLAHOE-UHFFFAOYSA-N
2,4,6,8-tetramethyl-2,4,6,8-tetravinylcyclotetrasiloxane,2,4,6,8-tetramethyl-2,4,6,8-tetravinyl-1,3,5,7,2,4,6,8-tetraoxatetrasilocane,cyclotetrasiloxane, 2,4,6,8-tetraethenyl-2,4,6,8-tetramethyl,1,3,5,7-tetravinyl-1,3,5,7-tetramethylcyclotetrasiloxane,1,3,5,7-tetramethyl-1,3,5,7-tetravinylcyclotetrasiloxane,2,4,6,8-tetravinyl-2,4,6,8-tetramethylcyclotetrasiloxane,tetravinyltetramethylcyclotetrasiloxane,s-tetramethyltetravinylcyclotetrasiloxane,unii-3nt606t1vk,cyclotetrasiloxane, tetraethenyltetramethyl
75706
2,4,6,8-tetrakis(ethenyl)-2,4,6,8-tetramethyl-1,3,5,7,2,4,6,8-tetraoxatetrasilocane
C[Si]1(O[Si](O[Si](O[Si](O1)(C)C=C)(C)C=C)(C)C=C)C=C
2550-06-3
C12H24O4Si4
344.66
MFCD00013694
VMAWODUEPLAHOE-UHFFFAOYSA-N
2,4,6,8-tetramethyl-2,4,6,8-tetravinylcyclotetrasiloxane,2,4,6,8-tetramethyl-2,4,6,8-tetravinyl-1,3,5,7,2,4,6,8-tetraoxatetrasilocane,cyclotetrasiloxane, 2,4,6,8-tetraethenyl-2,4,6,8-tetramethyl,1,3,5,7-tetravinyl-1,3,5,7-tetramethylcyclotetrasiloxane,1,3,5,7-tetramethyl-1,3,5,7-tetravinylcyclotetrasiloxane,2,4,6,8-tetravinyl-2,4,6,8-tetramethylcyclotetrasiloxane,tetravinyltetramethylcyclotetrasiloxane,s-tetramethyltetravinylcyclotetrasiloxane,unii-3nt606t1vk,cyclotetrasiloxane, tetraethenyltetramethyl
75706
2,4,6,8-tetrakis(ethenyl)-2,4,6,8-tetramethyl-1,3,5,7,2,4,6,8-tetraoxatetrasilocane
C[Si]1(O[Si](O[Si](O[Si](O1)(C)C=C)(C)C=C)(C)C=C)C=C
Linear Formula Cl(CH2)3SiCl3
UN Number UN2987
Beilstein 1737851
Formula Weight 211.98
Percent Purity 97%
Density 1.36
Odor Acrid
Refractive Index 1.4675
Sensitivity Moisture sensitive
Boiling Point 181°C to 183°C
Quantity 100g
Flash Point 84°C (183°F)
Chemical Name or Material (3-Chloropropyl)trichlorosilane
Show More Show Less

GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.

P210-P235-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c

H227-H302-H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CHLOROSILANES, CORROSIVE, N.O.S.

EINECSNumber : 219-844-8

TSCA : Yes

Recommended Storage : Ambient temperatures