Alfa Aesar™ (3-Chloropropyl)trimethoxysilane, 97%

Manufacturer: Alfa Aesar™ B2251722

Quantity 100g

 View more versions of this product

Catalog No. AAB2251722

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 2530-87-2
    Molecular Formula ClCH2CH2CH2Si(OCH3)3
    Formula Weight 198.72
    MDL Number MFCD00000997
    Beilstein 969627
    Chemical Name or Material (3-Chloropropyl)trimethoxysilane

    Specifications

    Formula Weight 198.72
    TSCA TSCA
    Odor Recognizable
    Molecular Formula ClCH2CH2CH2Si(OCH3)3
    MDL Number MFCD00000997
    Boiling Point 195° to 197°C
    Melting Point −50°C
    Quantity 100g
    Flash Point 78°C (172°F)
    Health Hazard 3 GHS P Statement
    P280g-P305+P351+P338
    Wear protective gloves.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    RTECS Number VV2680000
    CAS 2530-87-2
    Beilstein 969627
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H315-H319-H335-H227
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Combustible liquid.
    Chemical Name or Material (3-Chloropropyl)trimethoxysilane
    Assay 97%

    Description

    Applications
    (3-Chloropropyl)trimethoxysilane is a good coupling agent for epoxy resins to glass in laminates.

    Solubility
    Hydrolyzes in water.

    Notes
    Moisture sensitive. Store away from strong oxidizing agents, water/ moisture.

    GHS H Statement
    H315-H319-H335-H227
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Combustible liquid.

    GHS P Statement
    P280g-P305+P351+P338
    Wear protective gloves.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit