Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(3-chloropropyl)trimethoxysilane, 98+%, ACROS Organics™

Click to view available options
Quantity:
25mL
100mL
500mL
Packaging:
Glass bottle
2530-87-2
C6H15ClO3Si
198.72
MFCD00000997
OXYZDRAJMHGSMW-UHFFFAOYSA-N
3-chloropropyl trimethoxysilane,silane, 3-chloropropyl trimethoxy,3-chloropropyltrimethyoxysilane,sila-ace s 620,cps-m,silane 3-chloropropyl tris methoxy,3-trimethoxysilyl propyl chloride,unii-t21bnl1s7f,cptmo,3-chloropropyl trimethoxysilan
62449
3-chloropropyl(trimethoxy)silane
CO[Si](CCCCl)(OC)OC
2530-87-2
C6H15ClO3Si
198.72
MFCD00000997
OXYZDRAJMHGSMW-UHFFFAOYSA-N
3-chloropropyl trimethoxysilane,silane, 3-chloropropyl trimethoxy,3-chloropropyltrimethyoxysilane,sila-ace s 620,cps-m,silane 3-chloropropyl tris methoxy,3-trimethoxysilyl propyl chloride,unii-t21bnl1s7f,cptmo,3-chloropropyl trimethoxysilan
62449
3-chloropropyl(trimethoxy)silane
CO[Si](CCCCl)(OC)OC
Flash Point 78°C
Boiling Point 195°C to 196°C (750.0mmHg)
Density 1.0700g/mL
Specific Gravity 1.07
Refractive Index 1.4190 to 1.421
Packaging Glass bottle
Quantity 100mL
Infrared Spectrum Authentic
Color Colorless to Yellow
CAS Min % 98.0
CAS Max % 100.0
Assay Percent Range 98+%
Linear Formula (CH3O)3Si(CH2)3Cl
Solubility Information Solubility in water: soluble
Formula Weight 198.72
Physical Form Liquid
Percent Purity ≥98.0%
Chemical Name or Material (3-Chloropropyl)trimethoxysilane
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning